4-Bromo-2-chlorotoluene ≥98%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 1-Bromo-3-chloro-4-methylbenzene

B22293.14 B22293.22
ALFAB22293.14EA 0 SGD
ALFAB22293.14 ALFAB22293.22
4-Bromo-2-chlorotoluene ≥98%
4-Bromo-2-chlorotoluene

Purity: 98% CAS No.: 89794-02-5 Molecular Formula: C7H6BrCl Formula Weight: 205.48 Flash Point: 104°(219°F) Refractive Index: 1.5735 Boiling Point: 212° Beilstein Reference: 3234853 MDL No.: MFCD00060651

Formula: C₇H₆BrCl
Boiling Pt: 212 °C
Density: 1,538
Flash Pt: 104 °C (219 °F)
Storage Temperature: Ambient
MDL Number: MFCD00060651
CAS Number: 89794-02-5

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR