2-Bromo-5-iodotoluene ≥98%

Supplier: Thermo Fisher Scientific
Warning

B22462.09 B22462.88
ALFAB22462.09EA 0 SGD
ALFAB22462.09 ALFAB22462.88
2-Bromo-5-iodotoluene ≥98%
2-Bromo-5-iodotoluene

Purity: 98+% CAS No.: 202865-85-8 Molecular Formula: C7H6BrI Formula Weight: 296.93 Refractive Index: 1.6490 Beilstein Reference: 8760525

Formula: C₇H₆BrI
Storage Temperature: Ambient
MDL Number: MFCD00070749
CAS Number: 202865-85-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR