N-Fmoc-L-norleucine 98

Supplier: Thermo Fisher Scientific

Synonyms: Fmoc-Nle-OH, FMOC-L-Norleucine

B22475.03 B22475.06 B22475.14
ALFAB22475.03EA 0 SGD
ALFAB22475.03 ALFAB22475.06 ALFAB22475.14
N-Fmoc-L-norleucine 98
N-Fmoc-L-norleucine

Purity: 98% CAS No.: 77284-32-3 Molecular Formula: C21H23NO4 Formula Weight: 353.42 Storage Temperature: 2°C to 8°C Melting Point: 140-142° MDL No.: MFCD00037537

Formula: C₂₁H₂₃NO₄
MW: 353.4 g/mol
Melting Pt: 140…142 °C
MDL Number: MFCD00037537
CAS Number: 77284-32-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR