2,3,4,5,6-Pentamethylbenzyl alcohol ≥96%

Supplier: Thermo Fisher Scientific

B22574.03
ALFAB22574.03EA 0 SGD
ALFAB22574.03
2,3,4,5,6-Pentamethylbenzyl alcohol ≥96%
2,3,4,5,6-Pentamethylbenzyl alcohol

Purity: 96% CAS No.: 484-66-2 Molecular Formula: C12H18O Formula Weight: 178.28 Melting Point: 158-160° Beilstein Reference: 2208765

Formula: C₁₂H₁₈O
Melting Pt: 158…160 °C
Storage Temperature: Ambient
MDL Number: MFCD00014421
CAS Number: 484-66-2
EINECS: 207-609-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR