Ethyl 2-bromoisobutyrate ≥98%

Supplier: Thermo Fisher Scientific
Danger

B22785.18 B22785.30
ALFAB22785.18EA 0 SGD
ALFAB22785.18 ALFAB22785.30
Ethyl 2-bromoisobutyrate ≥98%
Ethyl 2-bromoisobutyrate

Purity: 98+% CAS No.: 600-00-0 Molecular Formula: C6H11BrO2 Formula Weight: 195.06 Flash Point: 57°(135°F) Refractive Index: 1.4440 Boiling Point: 162-164° Beilstein Reference: 1098947 UN No.: 2924 MDL No.: MFCD00000123

Formula: C₆H₁₁BrO₂
Boiling Pt: 162…164 °C
Density: 1,329
Flash Pt: 57 °C (135 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000123
CAS Number: 600-00-0
EINECS: 209-980-6
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR