(R)-(-)-2,2-Dimethyl-1,3-dioxolan-4-ylmethanol 96%

Supplier: Thermo Fisher Scientific
Warning

B23037.03 B23037.06
ALFAB23037.03EA 0 SGD
ALFAB23037.03 ALFAB23037.06
(R)-(-)-2,2-Dimethyl-1,3-dioxolan-4-ylmethanol 96%
(R)-(-)-2,2-Dimethyl-1,3-dioxolan-4-ylmethanol

Purity: 96% CAS No.: 14347-78-5 Molecular Formula: C6H12O3 Formula Weight: 132.18 Storage Temperature: 2°C to 8°C Flash Point: 80°(176°F) Refractive Index: 1.4350 Boiling Point: 82-83°/14mm Beilstein Reference: 80117 Merck Reference: 14,5213 Optical Rotation: -11.5° (c=5 in methanol) MDL No.: MFCD00003213

Formula: C₆H₁₂O₃
Boiling Pt: 82…83 °C (14 mmHg)
Density: 1,066
Flash Pt: 80 °C (176 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00003213
CAS Number: 14347-78-5
EINECS: 405-320-8
Merck Index: 14,5213

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR