3-Fluoro-α-methylbenzyl alcohol ≥97%

Supplier: Thermo Fisher Scientific
Warning

B23144.06 B23144.14
ALFAB23144.06EA 0 SGD
ALFAB23144.06 ALFAB23144.14
3-Fluoro-α-methylbenzyl alcohol ≥97%
3-Fluoro-α-methylbenzyl alcohol

Purity: 97% CAS No.: 402-63-1 Molecular Formula: C8H9FO Formula Weight: 140.16 Flash Point: 93°(199°F) Refractive Index: 1.5035 Boiling Point: 104-106°/20mm Beilstein Reference: 1932847 MDL No.: MFCD00014402

Formula: C₈H₉FO
Boiling Pt: 104…106 °C (20 mmHg)
Density: 1,210
Flash Pt: 93 °C (199 °F)
Storage Temperature: Ambient
MDL Number: MFCD00014402
CAS Number: 402-63-1
EINECS: 206-950-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR