3-(Hydroxymethyl)benzyl alcohol ≥98%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 1,3-Benzenedimethanol, m-Xylene-α,α'-diol, m-Phenylenedimethanol

B23417.03 B23417.06 B23417.14
ALFAB23417.03EA 0 SGD
ALFAB23417.03 ALFAB23417.06 ALFAB23417.14
3-(Hydroxymethyl)benzyl alcohol ≥98%
3-(Hydroxymethyl)benzyl alcohol

Purity: 98% CAS No.: 626-18-6 Molecular Formula: C8H10O2 Formula Weight: 138.17 Melting Point: 56-60° Boiling Point: 154-159°/13mm MDL No.: MFCD00004649

Formula: C₈H₁₀O₂
Boiling Pt: 154…159 °C (13 mmHg)
Melting Pt: 56…60 °C
Density: 1,161 g/cm³ (18 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004649
CAS Number: 626-18-6
EINECS: 210-934-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR