4-Amino-2-chlorophenol 99%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: 3-Chloro-4-hydroxyaniline

B23420.03 B23420.06 B23420.14
ALFAB23420.03EA 0 SGD
ALFAB23420.03 ALFAB23420.06 ALFAB23420.14
4-Amino-2-chlorophenol 99%
4-Amino-2-chlorophenol

Purity: 99% CAS No.: 3964-52-1 Molecular Formula: C6H6ClNO Formula Weight: 143.57 Melting Point: 150-153° MDL No.: MFCD00128891

Formula: C₆H₆ClNO
Melting Pt: 150…153 °C
Storage Temperature: Ambient
MDL Number: MFCD00128891
CAS Number: 3964-52-1

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR