4'-Bromo-4-chlorobutyrophenone ≥98%

Supplier: Thermo Fisher Scientific

B24146.14 B24146.22
ALFAB24146.14EA 0 SGD
ALFAB24146.14 ALFAB24146.22
4'-Bromo-4-chlorobutyrophenone ≥98%
4'-Bromo-4-chlorobutyrophenone

Purity: 98% CAS No.: 4559-96-0 Molecular Formula: C10H10BrClO Formula Weight: 261.55 Flash Point: 110°(230°F) Melting Point: 35-38° MDL No.: MFCD00001006

Formula: C₁₀H₁₀BrClO
Boiling Pt: 135…139 °C (0,3 mmHg)
Melting Pt: 35…38 °C
Density: 1,4808 g/cm³ (20 °C)
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001006
CAS Number: 4559-96-0
EINECS: 224-930-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR