Ethyl iodoacetate ≥98%

Supplier: Thermo Fisher Scientific
Danger

B24177.06 B24177.14 B24177.22
ALFAB24177.06EA 0 SGD
ALFAB24177.06 ALFAB24177.14 ALFAB24177.22
Ethyl iodoacetate ≥98%
Ethyl iodoacetate

Purity: 98% CAS No.: 623-48-3 Molecular Formula: ICH2CO2CH2CH3 Formula Weight: 214 Storage Temperature: 2°C to 8°C Flash Point: 76°(169°F) Refractive Index: 1.503 Boiling Point: 179-180° Beilstein Reference: 741934 UN No.: 2927 MDL No.: MFCD00001081

Formula: C₄H₇IO₂
Boiling Pt: 179…180 °C
Density: 1,81
Flash Pt: 76 °C (169 °F)
MDL Number: MFCD00001081
CAS Number: 623-48-3
EINECS: 210-796-3
UN: 2927
ADR: 6.1,II

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR