ω-Bromo-4-phenylacetophenone 97

Supplier: Thermo Fisher Scientific
Danger

Synonyms: 2-Bromo-4'-phenylacetophenone, Bromomethyl p-phenylphenyl ketone, α-Bromo-4-phenylacetophenone, Biphenyl-4-yl bromomethyl ketone, 4-Phenylphenacyl bromide

B24298.06 B24298.14 B24298.22
ALFAB24298.06EA 0 SGD
ALFAB24298.06 ALFAB24298.14 ALFAB24298.22
ω-Bromo-4-phenylacetophenone 97
ω-Bromo-4-phenylacetophenone

Purity: 97% CAS No.: 135-73-9 Molecular Formula: C14H11BrO Formula Weight: 275.15 Melting Point: 123-127° Beilstein Reference: 513838 UN No.: 3261 MDL No.: MFCD00000202

Formula: C₁₄H₁₁BrO
MW: 275.14 g/mol
Boiling Pt: 354
Melting Pt: 123…127 °C
Storage Temperature: Ambient
MDL Number: MFCD00000202
CAS Number: 135-73-9
EINECS: 205-217-6
UN: 3261
ADR: 8 III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR