4'-Bromo-3'-nitroacetophenone ≥99%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 1-(4-Bromo-3-nitrophenyl)ethan-1-one, 1-(4-Bromo-3-nitrophenyl)ethanone

B24469.06 B24469.14
ALFAB24469.06EA 0 SGD
ALFAB24469.06 ALFAB24469.14
4'-Bromo-3'-nitroacetophenone ≥99%
4'-Bromo-3'-nitroacetophenone

Purity: 99% CAS No.: 18640-58-9 Molecular Formula: C8H6BrNO3 Formula Weight: 244.05 Melting Point: 117-120° Beilstein Reference: 2050098

Formula: C₈H₆BrNO₃
Melting Pt: 117…120 °C
Storage Temperature: Ambient
MDL Number: MFCD00016985
CAS Number: 18640-58-9
EINECS: 242-469-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR