(S)-(+)-2,2-Dimethyl-1,3-dioxolan-4-ylmethanol ≥98%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: (S)-(+)-1,2-O-Isopropylidenglycerol, (S)-(+)-2,2-Dimethyl-1,3-dioxolane-4-methanol, (S)-(+)-1,2-Isopropylideneglycerol, (+)-Solketal, (S)-(+)-2,3-O-Isopropylideneglycerol

B24488.04 B24488.09
ALFAB24488.04EA 0 SGD
ALFAB24488.04 ALFAB24488.09
(S)-(+)-2,2-Dimethyl-1,3-dioxolan-4-ylmethanol ≥98%
(S)-(+)-2,2-Dimethyl-1,3-dioxolan-4-ylmethanol

Purity: 98% CAS No.: 22323-82-6 Molecular Formula: C6H12O3 Formula Weight: 132.18 Storage Temperature: 2°C to 8°C Flash Point: 80°(176°F) Refractive Index: 1.4350 Boiling Point: 82-83°/14mm Beilstein Reference: 80118 Merck Reference: 14,5213 Optical Rotation: +11.5° (c=5 in methanol) MDL No.: MFCD00063239

Formula: C₆H₁₂O₃
Boiling Pt: 82…83 °C (14 mmHg)
Density: 1,066
Flash Pt: 80 °C (176 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00063239
CAS Number: 22323-82-6
EINECS: 244-910-8
Merck Index: 14,5213

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR