1-(Trimethylsilyl)-1-propyne ≥98%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Trimethyl-1-propynylsilane

B24875.03 B24875.06 B24875.14
ALFAB24875.03EA 0 SGD
ALFAB24875.03 ALFAB24875.06 ALFAB24875.14
1-(Trimethylsilyl)-1-propyne ≥98%
1-(Trimethylsilyl)-1-propyne

Purity: 98% CAS No.: 6224-91-5 Molecular Formula: CH3C≡CSi(CH3)3 Formula Weight: 112.25 Flash Point: -3°(26°F) Refractive Index: 1.4200 Melting Point: -69° Boiling Point: 99-100° Beilstein Reference: 1071311 UN No.: 1993 MDL No.: MFCD00009271

Formula: C₆H₁₂Si
MW: 112.25 g/mol
Boiling Pt: 99…100 °C
Melting Pt: –69 °C
Density: 0.753
Flash Pt: –3 °C (26 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009271
CAS Number: 6224-91-5
EINECS: 228-314-5
ADR: 3 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR