5-Bromo-2-iodobenzonitrile ≥98%

Supplier: Thermo Fisher Scientific
Danger

B24880.06 B24880.14
ALFAB24880.06EA 0 SGD
ALFAB24880.06 ALFAB24880.14
5-Bromo-2-iodobenzonitrile ≥98%
5-Bromo-2-iodobenzonitrile

Purity: 98+% CAS No.: 121554-10-7 Molecular Formula: C7H3BrIN Formula Weight: 307.92 Melting Point: 120-122° Beilstein Reference: 4860282 UN No.: 3276 MDL No.: MFCD00672950

Formula: C₇H₃BrIN
MW: 307.92 g/mol
Melting Pt: 120…122 °C
Storage Temperature: Ambient
MDL Number: MFCD00672950
CAS Number: 121554-10-7
EINECS: 601-788-4
UN: 3439
ADR: 6.1 III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR