Coniferyl alcohol 98

Supplier: Thermo Fisher Scientific

Synonyms: 4-(3-Hydroxyprop-1-en-1-yl)-2-methoxyphenol

B24949.01 B24949.03 B24949.06
ALFAB24949.01EA 0 SGD
ALFAB24949.01 ALFAB24949.03 ALFAB24949.06
Coniferyl alcohol 98
Coniferyl alcohol

Purity: 98% CAS No.: 458-35-5 Molecular Formula: C10H12O3 Formula Weight: 180.20 Storage Temperature: -30°C to -10°C Melting Point: 75-76° Boiling Point: 163-165°/3mm Beilstein Reference: 2048961 Merck Reference: 14,2504 MDL No.: MFCD00002922

Formula: C₁₀H₁₂O₃
MW: 180.20 g/mol
Boiling Pt: 163…165 °C (3 mmHg)
Melting Pt: 75…76 °C
MDL Number: MFCD00002922
CAS Number: 458-35-5
EINECS: 207-277-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR