2-Bromo-5-methylpyridine ≥98%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: 2-Bromo-5-picoline

B25083.06 B25083.14 B25083.22
ALFAB25083.06EA 0 SGD
ALFAB25083.06 ALFAB25083.14 ALFAB25083.22
2-Bromo-5-methylpyridine ≥98%
2-Bromo-5-methylpyridine

Purity: 98+% CAS No.: 3510-66-5 Molecular Formula: C6H6BrN Formula Weight: 172.03 Flash Point: 103°(217°F) Melting Point: 40-45° Boiling Point: 95-96°/12mm Beilstein Reference: 107323 MDL No.: MFCD00209553

Formula: C₆H₆BrN
Boiling Pt: 95…96 °C (12 mmHg)
Melting Pt: 40…45 °C
Flash Pt: 103 °C (217 °F)
Storage Temperature: Ambient
MDL Number: MFCD00209553
CAS Number: 3510-66-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR