2-Ethyl-6-methylphenol ≥97%

Supplier: Thermo Fisher Scientific
Warning

B25213.03 B25213.06 B25213.14
ALFAB25213.03EA 0 SGD
ALFAB25213.03 ALFAB25213.06 ALFAB25213.14
2-Ethyl-6-methylphenol ≥97%
2-Ethyl-6-methylphenol

Purity: 97% CAS No.: 1687-64-5 Molecular Formula: C9H12O Formula Weight: 136.19 Refractive Index: 1.5310

Formula: C₉H₁₂O
Storage Temperature: Ambient
MDL Number: MFCD00962292
CAS Number: 1687-64-5

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR