Propyl hexanoate 98

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Hexanoic acid propyl ester, n-Propyl hexanoate, Propyl caproate, Caproic acid propyl ester

B25215.36
ALFAB25215.36EA 0 SGD
ALFAB25215.36
Propyl hexanoate 98
Propyl hexanoate

Purity: 98% CAS No.: 626-77-7 Molecular Formula: C9H18O2 Formula Weight: 158.24 Flash Point: 52°(126°F) Refractive Index: 1.4120 Odour: Fruit-like Melting Point: -69° Boiling Point: 187° UN No.: 3272 MDL No.: MFCD00053803

Formula: C₉H₁₈O₂
MW: 158.24 g/mol
Boiling Pt: 187 °C
Melting Pt: –69 °C
Density: 0.876
Flash Pt: 52 °C (126 °F)
Storage Temperature: Ambient
MDL Number: MFCD00053803
CAS Number: 626-77-7
EINECS: 210-963-0
UN: 3272
ADR: 3 III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR