1-Bromo-4-phenylbutane 98

Supplier: Thermo Fisher Scientific

B25237.03 B25237.06
ALFAB25237.03EA 81.6 SGD
ALFAB25237.03 ALFAB25237.06
1-Bromo-4-phenylbutane 98
1-Bromo-4-phenylbutane

Purity: 98% CAS No.: 13633-25-5 Molecular Formula: C10H13Br Formula Weight: 213.12 Boiling Point: 131-133°/12mm MDL No.: MFCD00154988

Formula: C₁₀H₁₃Br
MW: 213.12
Boiling Pt: 131…133 °C (12 mmHg)
Density: 1.214
Storage Temperature: Ambient
MDL Number: MFCD00154988
CAS Number: 13633-25-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR