Sodium alginate, low viscosity

Supplier: Thermo Fisher Scientific

Synonyms: Alginic acid sodium salt, Algin

B25266.22 B25266.0E B25266.36
ALFAB25266.22EA 89.4 SGD
ALFAB25266.22 ALFAB25266.0E ALFAB25266.36
Sodium alginate, low viscosity
Sodium alginate

CAS No.: 9005-38-3 Merck Reference: 14,241 MDL No.: MFCD00081310

Storage Temperature: Ambient
MDL Number: MFCD00081310
CAS Number: 9005-38-3
Merck Index: 14,241

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR