4-Bromo-2-methylphenyl isothiocyanate 98

Supplier: Thermo Fisher Scientific
Danger

B25296.06 B25296.14
ALFAB25296.06EA 0 SGD
ALFAB25296.06 ALFAB25296.14
4-Bromo-2-methylphenyl isothiocyanate 98
4-Bromo-2-methylphenyl isothiocyanate

Purity: 98% CAS No.: 19241-38-4 Molecular Formula: C8H6BrNS Formula Weight: 228.12 Melting Point: 44-46° Boiling Point: 129-130°/3mm UN No.: 2923 MDL No.: MFCD00041087

Formula: C₈H₆BrNS
MW: 228.11 g/mol
Boiling Pt: 129…130 °C (3 mmHg)
Melting Pt: 44…46 °C
Storage Temperature: Ambient
MDL Number: MFCD00041087
CAS Number: 19241-38-4
EINECS: 242-907-6
UN: 2923
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR