2-Bromo-4-chlorotoluene ≥98%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 1-Bromo-3-chloro-6-methylbenzene, 2-Bromo-4-chloro-1-methylbenzene

B25329.14 B25329.22 B25329.06
ALFAB25329.14EA 0 SGD
ALFAB25329.14 ALFAB25329.22 ALFAB25329.06
2-Bromo-4-chlorotoluene ≥98%
2-Bromo-4-chlorotoluene

Purity: 98% CAS No.: 27139-97-5 Molecular Formula: C7H6BrCl Formula Weight: 205.48 MDL No.: MFCD00060650

Formula: C₇H₆BrCl
MW: 205.48 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00060650
CAS Number: 27139-97-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR