3-Amino-4-methylbenzamide 97%

Supplier: Thermo Fisher Scientific
Warning

B25367.06
ALFAB25367.06EA 0 SGD
ALFAB25367.06
3-Amino-4-methylbenzamide 97%
3-Amino-4-methylbenzamide

Purity: 97% CAS No.: 19406-86-1 Molecular Formula: C8H10N2O Formula Weight: 150.18 Melting Point: 127-131° MDL No.: MFCD00035936

Formula: C₈H₁₀N₂O
MW: 150.18 g/mol
Melting Pt: 127…131 °C
Storage Temperature: Ambient
MDL Number: MFCD00035936
CAS Number: 19406-86-1
EINECS: 243-039-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR