2-Bromo-4-nitrophenetole 99%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: 3-Bromo-4-ethoxynitrobenzene

B25433.22 B25433.06
ALFAB25433.22EA 0 SGD
ALFAB25433.22 ALFAB25433.06
2-Bromo-4-nitrophenetole 99%
2-Bromo-4-nitrophenetole

Purity: 99% CAS No.: 58244-42-1 Molecular Formula: C8H8BrNO3 Formula Weight: 246.06 Melting Point: 94-98° MDL No.: MFCD12547883

Formula: C₈H₈BrNO₃
MW: 246.06 g/mol
Melting Pt: 94…98 °C
Storage Temperature: Ambient
MDL Number: MFCD12547883
CAS Number: 58244-42-1

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR