4'-(4-Methoxyphenyl)-2,2':6',2''-terpyridine 98%

Supplier: Thermo Fisher Scientific
Warning

B25486.02 B25486.77
ALFAB25486.02EA 0 SGD
ALFAB25486.02 ALFAB25486.77
4'-(4-Methoxyphenyl)-2,2':6',2''-terpyridine 98%
4'-(4-Methoxyphenyl)-2,2':6',2''-terpyridine

Purity: 98% CAS No.: 13104-56-8 Molecular Formula: C22H17N3O Formula Weight: 339.40 Melting Point: 161-163° MDL No.: MFCD06796987

Formula: C₂₂H₁₇N₃O
MW: 339.40 g/mol
Melting Pt: 161…163 °C
Storage Temperature: Ambient
MDL Number: MFCD06796987
CAS Number: 13104-56-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR