5-Bromo-2-hydroxybenzophenone ≥97%

Supplier: Thermo Fisher Scientific
Warning

B25507.06 B25507.14
ALFAB25507.06EA 240 SGD
ALFAB25507.06 ALFAB25507.14
5-Bromo-2-hydroxybenzophenone ≥97%
5-Bromo-2-hydroxybenzophenone

Purity: 97% CAS No.: 55082-33-2 Molecular Formula: C13H9BrO2 Formula Weight: 277.11 Melting Point: 108-111° MDL No.: MFCD00525062

Formula: C₁₃H₉BrO₂
Melting Pt: 108…111 °C
Storage Temperature: Ambient
MDL Number: MFCD00525062
CAS Number: 55082-33-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR