(±)-Ethyl-2-bromovalerate ≥97%

Supplier: Thermo Fisher Scientific
Danger

B25530.14 B25530.22
ALFAB25530.14EA 0 SGD
ALFAB25530.14 ALFAB25530.22
(±)-Ethyl-2-bromovalerate ≥97%
(±)-Ethyl 2-bromovalerate

Purity: 97% CAS No.: 615-83-8 Molecular Formula: C7H13BrO2 Formula Weight: 209.09 Flash Point: 77°(170°F) Refractive Index: 1.4480 Boiling Point: 190-192° Beilstein Reference: 1098901 UN No.: 3265 MDL No.: MFCD00000159

Formula: C₇H₁₃BrO₂
Boiling Pt: 190…192 °C
Density: 1,226
Flash Pt: 77 °C (170 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000159
CAS Number: 615-83-8
EINECS: 210-450-1
UN: 3265
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR