2-Bromo-6-methoxyphenol ≥98%

Supplier: Thermo Fisher Scientific
Warning

B25535.03 B25535.06 B25535.14
ALFAB25535.03EA 0 SGD
ALFAB25535.03 ALFAB25535.06 ALFAB25535.14
2-Bromo-6-methoxyphenol ≥98%
2-Bromo-6-methoxyphenol

Purity: 98+% CAS No.: 28165-49-3 Molecular Formula: C7H7BrO2 Formula Weight: 203.04 Melting Point: 62-65° MDL No.: MFCD08146628

Formula: C₇H₇BrO₂
MW: 203.04 g/mol
Melting Pt: 62…65 °C
Storage Temperature: Ambient
MDL Number: MFCD08146628
CAS Number: 28165-49-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR