4-Bromo-m-anisidine ≥97%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: 4-Bromo-3-methoxyaniline, 5-Amino-2-bromoanisole, 4-Bromo-3-anisidine

B25541.06 B25541.14
ALFAB25541.06EA 0 SGD
ALFAB25541.06 ALFAB25541.14
4-Bromo-m-anisidine ≥97%
4-Bromo-m-anisidine

Purity: 97+% CAS No.: 19056-40-7 Molecular Formula: C7H8BrNO Formula Weight: 202.05 Melting Point: 96-100° Beilstein Reference: 2690835 UN No.: 2811 MDL No.: MFCD05664063

Formula: C₇H₈BrNO
MW: 202.05 g/mol
Melting Pt: 96…100 °C
Storage Temperature: Ambient
MDL Number: MFCD05664063
CAS Number: 19056-40-7
UN: 2811
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR