2-Bromo-5-nitrobenzoic acid ≥98%

Supplier: Thermo Fisher Scientific
Warning

B25704.06
ALFAB25704.06EA 0 SGD
ALFAB25704.06
2-Bromo-5-nitrobenzoic acid ≥98%
2-Bromo-5-nitrobenzoic acid

Purity: 98% CAS No.: 943-14-6 Molecular Formula: C7H4BrNO4 Formula Weight: 246.02 Melting Point: 178-181° Beilstein Reference: 980242

Formula: C₇H₄BrNO₄
MW: 246.02 g/mol
Boiling Pt: 371
Melting Pt: 178…181 °C
Storage Temperature: Ambient
MDL Number: MFCD00134558
CAS Number: 943-14-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR