Ethyl-5-bromo-1,2,3-thiadiazole-4-carboxylate 98%

Supplier: Thermo Fisher Scientific
Warning

H25764.03
ALFAH25764.03EA 0 SGD
ALFAH25764.03
Ethyl-5-bromo-1,2,3-thiadiazole-4-carboxylate 98%
Ethyl-5-bromo-1,2,3-thiadiazole-4-carboxylate

Purity: 98% CAS No.: 6439-91-4 Molecular Formula: C5H5BrN2O2S Formula Weight: 237.07 Melting Point: 76-78° MDL No.: MFCD02324685

Formula: C₅H₅BrN₂O₂S
MW: 237.07 g/mol
Melting Pt: 76…78 °C
Storage Temperature: Ambient
MDL Number: MFCD02324685
CAS Number: 6439-91-4

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR