2-Bromo-6-fluoroaniline ≥98%

Supplier: Thermo Fisher Scientific
Danger

H25774.06 H25774.14
ALFAH25774.06EA 0 SGD
ALFAH25774.06 ALFAH25774.14
2-Bromo-6-fluoroaniline ≥98%
2-Bromo-6-fluoroaniline

Purity: 98+% CAS No.: 65896-11-9 Molecular Formula: C6H5BrFN Formula Weight: 190.01 Refractive Index: 1.5820 Beilstein Reference: 8541957 UN No.: 2810 MDL No.: MFCD01310982

Formula: C₆H₅BrFN
MW: 190.01 g/mol
Density: 1.674
Storage Temperature: Ambient
MDL Number: MFCD01310982
CAS Number: 65896-11-9
UN: 2810
ADR: 6.1 III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR