2-Bromo-4-chloro-1-fluorobenzene ≥97%

Supplier: Thermo Fisher Scientific
Warning

H25935.14 H25935.06
ALFAH25935.14EA 0 SGD
ALFAH25935.14 ALFAH25935.06
2-Bromo-4-chloro-1-fluorobenzene ≥97%
2-Bromo-4-chloro-1-fluorobenzene

Purity: 97% CAS No.: 1996-30-1 Molecular Formula: C6H3BrClF Formula Weight: 209.45 Flash Point: 85°(185°F) Refractive Index: 1.554 Boiling Point: 180° MDL No.: MFCD00672934

Formula: C₆H₃BrClF
Boiling Pt: 180 °C
Density: 1,74
Flash Pt: 85 °C (185 °F)
Storage Temperature: Ambient
MDL Number: MFCD00672934
CAS Number: 1996-30-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR