3,4-Bis(trifluoromethyl)pyridine 96%

Supplier: Thermo Fisher Scientific
Warning

H26008.03
ALFAH26008.03EA 0 SGD
ALFAH26008.03
3,4-Bis(trifluoromethyl)pyridine 96%
3,4-Bis(trifluoromethyl)pyridine

Purity: 96% CAS No.: 20857-46-9 Molecular Formula: C7H3F6N Formula Weight: 215.10 Refractive Index: 1.3920 Boiling Point: 42-44°/24mm MDL No.: MFCD08686956

Formula: C₇H₃F₆N
Boiling Pt: 42…44 °C (24 mmHg)
Storage Temperature: Ambient
MDL Number: MFCD08686956
CAS Number: 20857-46-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR