Benzyl(4-oxocyclohexyl)carbamate 97%

Supplier: Thermo Fisher Scientific

Synonyms: Benzyl (4-oxocyclohexyl)carbamate, 4-N-Cbz-aminocyclohexanone

H26015.06
ALFAH26015.06EA 0 SGD
ALFAH26015.06
Benzyl(4-oxocyclohexyl)carbamate 97%
Benzyl(4-oxocyclohexyl)carbamate

Purity: 97% CAS No.: 16801-63-1 Molecular Formula: C14H17NO3 Formula Weight: 247.29 Odour: Odourless Melting Point: 81-83° MDL No.: MFCD06658393

Formula: C₁₄H₁₇NO₃
Melting Pt: 81…83 °C
Storage Temperature: Ambient
MDL Number: MFCD06658393
CAS Number: 16801-63-1

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR