2-Chloro-6-methylbenzaldehyde 98%

Supplier: Thermo Fisher Scientific
Warning

H26023.03 H26023.06
ALFAH26023.03EA 0 SGD
ALFAH26023.03 ALFAH26023.06
2-Chloro-6-methylbenzaldehyde 98%
2-Chloro-6-methylbenzaldehyde

Purity: 98% CAS No.: 1194-64-5 Molecular Formula: C8H7ClO Formula Weight: 154.59 Flash Point: 99°(210°F) Melting Point: 36-38° Boiling Point: 74°/0.4mm MDL No.: MFCD01934409

Formula: C₈H₇ClO
MW: 154.6 g/mol
Boiling Pt: 74 °C (0.4 mmHg)
Melting Pt: 36…38 °C
Flash Pt: 99 °C (210 °F)
Storage Temperature: Ambient
MDL Number: MFCD01934409
CAS Number: 1194-64-5

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR