4-Methyl-3-nitrobenzenesulphonamide 97%

Supplier: Thermo Fisher Scientific
Warning

H26039.03
ALFAH26039.03EA 0 SGD
ALFAH26039.03
4-Methyl-3-nitrobenzenesulphonamide 97%
4-Methyl-3-nitrobenzenesulphonamide

Purity: 97% CAS No.: 6949-23-1 Molecular Formula: C7H8N2O4S Formula Weight: 216.22 Melting Point: 141-144° MDL No.: MFCD00460611

Formula: C₇H₈N₂O₄S
MW: 216.22 g/mol
Melting Pt: 141…144 °C
Storage Temperature: Ambient
MDL Number: MFCD00460611
CAS Number: 6949-23-1
EINECS: 230-117-4

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR