2-(1,3-Dioxolan-2-yl)ethylboronic acid pinacol ester ≥97%

Supplier: Thermo Fisher Scientific

Synonyms: 2-(2-(1,3-Dioxolan-2-yl)ethyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

H26071.03 H26071.06
ALFAH26071.03EA 0 SGD
ALFAH26071.03 ALFAH26071.06
2-(1,3-Dioxolan-2-yl)ethylboronic acid pinacol ester ≥97%
2-(1,3-Dioxolan-2-yl)ethylboronic acid pinacol ester

Purity: 97% Molecular Formula: C11H21BO4 Formula Weight: 228.10 Boiling Point: 100°/1.5mm MDL No.: MFCD03788722

Formula: C₁₁H₂₁BO₄
MW: 228.10 g/mol
Boiling Pt: 100 °C (1.5 mmHg)
Storage Temperature: Ambient
MDL Number: MFCD03788722
CAS Number: 1073354-07-0

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR