Methyl-9-octadecynoate 90

Supplier: Thermo Fisher Scientific
Warning

H26080.03
ALFAH26080.03EA 0 SGD
ALFAH26080.03
Methyl-9-octadecynoate 90
Methyl-9-octadecynoate

Purity: 90+% CAS No.: 1120-32-7 Molecular Formula: CH3(CH2)7C≡C(CH2)7CO2CH3 Formula Weight: 294.47 MDL No.: MFCD09953468

Formula: CH₃(CH₂)₇CΞC(CH₂)₇CO₂CH₃
MW: 294.47 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09953468
CAS Number: 1120-32-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR