4-Bromo-3,5-difluorophenol ≥99%

Supplier: Thermo Fisher Scientific
Warning

H26153.03
ALFAH26153.03EA 0 SGD
ALFAH26153.03
4-Bromo-3,5-difluorophenol ≥99%
4-Bromo-3,5-difluorophenol

Purity: 99% CAS No.: 130191-91-2 Molecular Formula: C6H3BrF2O Formula Weight: 208.99 Melting Point: 72-78° MDL No.: MFCD08062385

Formula: C₆H₃BrF₂O
Melting Pt: 72…78 °C
Storage Temperature: Ambient
MDL Number: MFCD08062385
CAS Number: 130191-91-2
EINECS: None

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR