3-Bromo-2,6-diluorotoluene ≥99%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 3-Bromo-2,6-difluorotoluene, 1-Bromo-2,4-difluoro-3-methylbenzene

H26159.03
ALFAH26159.03EA 0 SGD
ALFAH26159.03
3-Bromo-2,6-diluorotoluene ≥99%
3-Bromo-2,6-diluorotoluene

Purity: 99% CAS No.: 221220-97-9 Molecular Formula: C7H5BrF2 Formula Weight: 207.02 Refractive Index: 1.5100

Formula: C₇H₅BrF₂
MW: 207.02 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03412221
CAS Number: 221220-97-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR