3-Bromo-α,α,α-trifluoro-5-nitrotoluene ≥97%

Supplier: Thermo Fisher Scientific

H26171.06
ALFAH26171.06EA 0 SGD
ALFAH26171.06
3-Bromo-α,α,α-trifluoro-5-nitrotoluene ≥97%
3-Bromo-α,α,α-trifluoro-5-nitrotoluene

Purity: 97% CAS No.: 630125-49-4 Molecular Formula: C7H3BrF3NO2 Formula Weight: 270.01 Refractive Index: 1.5140

Formula: C₇H₃BrF₃NO₂
MDL Number: MFCD03788719
CAS Number: 630125-49-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR