5-Amino-2-fluorobenzonitrile 97%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: 3-Cyano-4-fluoroaniline

H26210.06 H26210.14
ALFAH26210.06EA 0 SGD
ALFAH26210.06 ALFAH26210.14
5-Amino-2-fluorobenzonitrile 97%
5-Amino-2-fluorobenzonitrile

Purity: 97% CAS No.: 53312-81-5 Molecular Formula: C7H5FN2 Formula Weight: 136.13 Melting Point: 92-96° UN No.: 3439 MDL No.: MFCD00277872

Formula: C₇H₅FN₂
Melting Pt: 92…96 °C
Storage Temperature: Ambient
MDL Number: MFCD00277872
CAS Number: 53312-81-5
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR