Bis(toluene-4-sulphonyl)methane 97%

Supplier: Thermo Fisher Scientific

H26617.MD H26617.03 H26617.06
ALFAH26617.MDEA 0 SGD
ALFAH26617.MD ALFAH26617.03 ALFAH26617.06
Bis(toluene-4-sulphonyl)methane 97%
Bis(toluene-4-sulphonyl)methane

Purity: 97% CAS No.: 15310-28-8 Molecular Formula: C15H16O4S2 Formula Weight: 324.42 Melting Point: 137-139° MDL No.: MFCD00032255

Formula: C₁₅H₁₆O₄S₂
Melting Pt: 137…139 °C
Storage Temperature: Ambient
MDL Number: MFCD00032255
CAS Number: 15310-28-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR