Trimethylsilyl-2,2-difluoro-2-(fluorosulphonyl)acetate ≥94%

Supplier: Thermo Fisher Scientific
Danger

H26857.06 H26857.14
ALFAH26857.06EA 0 SGD
ALFAH26857.06 ALFAH26857.14
Trimethylsilyl-2,2-difluoro-2-(fluorosulphonyl)acetate ≥94%
Trimethylsilyl 2,2-difluoro-2-(fluorosulphonyl)acetate

Purity: 94% CAS No.: 120801-75-4 Molecular Formula: FSO2CF2CO2Si(CH3)3 Formula Weight: 250.27 Flash Point: 26°(79°F) Refractive Index: 1.367 Boiling Point: 156° UN No.: 2920 MDL No.: MFCD02093343

Formula: C₅H₉F₃O₄SSi
MW: 250.27 g/mol
Boiling Pt: 156 °C
Density: 1.27
Flash Pt: 26 °C (79 °F)
Storage Temperature: Ambient
MDL Number: MFCD02093343
CAS Number: 120801-75-4
ADR: 8 II

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR