5-Bromo-3-nitrosalicylaldehyde ≥97%

Supplier: Thermo Fisher Scientific
Warning

H26927.06 H26927.03
ALFAH26927.06EA 0 SGD
ALFAH26927.06 ALFAH26927.03
5-Bromo-3-nitrosalicylaldehyde ≥97%
5-Bromo-3-nitrosalicylaldehyde

Purity: 97% CAS No.: 16634-88-1 Molecular Formula: C7H4BrNO4 Formula Weight: 246.02 Melting Point: 125-128° MDL No.: MFCD00130110

Formula: C₇H₄BrNO₄
Melting Pt: 125…128 °C
Storage Temperature: Ambient
MDL Number: MFCD00130110
CAS Number: 16634-88-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR