5-Bromo-o-cresol 95%

Supplier: Thermo Fisher Scientific
Warning

H26956.09 H26956.03
ALFAH26956.09EA 0 SGD
ALFAH26956.09 ALFAH26956.03
5-Bromo-o-cresol 95%
5-Bromo-o-cresol

Purity: 95% CAS No.: 36138-76-8 Molecular Formula: C7H7BrO Formula Weight: 187.04 Melting Point: 77-80° MDL No.: MFCD08061906

Formula: C₇H₇BrO
Melting Pt: 77…80 °C
Storage Temperature: Ambient
MDL Number: MFCD08061906
CAS Number: 36138-76-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR