1-Butyl-3-methylpyridiniumtetrafluoroborate ≥99%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 1-Butyl-3-methylpyridin-1-ium tetrafluoroborate

H26983.18 H26983.06
ALFAH26983.18EA 0 SGD
ALFAH26983.18 ALFAH26983.06
1-Butyl-3-methylpyridiniumtetrafluoroborate ≥99%
1-Butyl-3-methylpyridinium tetrafluoroborate

Purity: 99% CAS No.: 597581-48-1 Molecular Formula: C10H16BF4N Formula Weight: 237.06 MDL No.: MFCD08458499

Formula: C₁₀H₁₆BF₄N
Density: 1,204
Storage Temperature: Ambient
MDL Number: MFCD08458499
CAS Number: 597581-48-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR